Assay Details
| Gene Symbol: Rpp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145938.4 | NP_666050.3 | 5 - 6 | 574 | 102 | |
| GenBank mRNA | AK048810.1 | - | 574 | 102 | ||
| BC024607.1 | - | 5 - 6 | 519 | 4236826 | 102 | |
| BC037666.1 | - | 3 - 4 | 2717 | 102 | ||
More Information
Set Membership: |
|

Rpp40
Genomic Map