Assay Details
| Gene Symbol: Omd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012050.2 | NP_036180.1 | 3 - 4 | 1198 | 106 | |
| GenBank mRNA | AB007848.1 | - | 3 - 4 | 1203 | 106 | |
| AK050297.1 | - | 1 - 2 | 301 | 106 | ||
| BC095997.1 | - | 2 - 3 | 1004 | 30739852 | 106 | |
More Information
Set Membership: |
|

Omd
Genomic Map