Assay Details
| Gene Symbol: Eef1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025380.2 | NP_079656.1 | 1 - 2 | 119 | 96 | |
| GenBank mRNA | AK003335.1 | - | 1 - 2 | 102 | 96 | |
| AK160723.1 | - | 1 - 2 | 102 | 96 | ||
| BC048466.1 | - | 1 - 2 | 101 | 6589105 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eef1e1
Genomic Map