Assay Details
| Gene Symbol: Eef1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025380.2 | NP_079656.1 | 2 - 3 | 322 | 106 | |
| GenBank mRNA | AK003335.1 | - | 2 - 3 | 305 | 106 | |
| AK009889.1 | - | 2 - 3 | 294 | 106 | ||
| AK199512.1 | - | 1 - 2 | 206 | 106 | ||
| BC048466.1 | - | 2 - 3 | 304 | 6589105 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eef1e1
Genomic Map