Assay Details
| Gene Symbol: Tfap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122948.2 | NP_001116420.1 | 1 - 2 | 418 | 69 | |
| GenBank mRNA | AK053704.1 | - | 1 - 2 | 403 | 69 | |
| U17287.1 | - | 1 - 2 | 382 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK053704.1, NM_001122948.2, U17287.1 transcripts.
Set Membership: |
|

Tfap2a
Genomic Map