Assay Details
| Gene Symbol: Hist3h2bb-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206882.1 | NP_996765.1 | 1 | 58 | 147 | |
| GenBank mRNA | BC107301.1 | - | 1 | 57 | 40053183 | 147 |
| BC107302.1 | - | 1 | 58 | 40053184 | 147 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107301.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27833725 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27833721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27833726 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27833720 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P108153 | NM_206882.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369321 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166682 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P108153 | NM_206882.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27833713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27833715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27833718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27833719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27833721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27833717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27833709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27833708 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166682 | - | - |
| Whole Mouse Genome G4122A | A_52_P527400 | NM_206882.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P527400 | BC107301.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P527400 | BC107302.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P527400 | NM_030082.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P527400 | AK018765.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108153 | NM_206882.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09483194 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09483217 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09483204 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27833726 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27833725 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27833721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27833720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27833696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27833708 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10386237 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376445 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10386236 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376444 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100025609.mm.1 | TC1100002837.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 70278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 70278 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070278 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257212 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257212 | - | - |
| mouse-6 v1.1 (Illumina) | scl0382522.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70278 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0382522.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70278 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257212 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257212 | - | - |

Hist3h2bb-ps
Genomic Map