Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_001166463.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_001166463.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_001166464.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_001166464.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_001166465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_001166465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_001166466.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_001166466.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_009262.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
NM_009262.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013247.mm.1 |
TC1300002086.mm.1 |
MTA TC |