Assay Details
| Gene Symbol: Pdlim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114087.2 | NP_001107559.1 | 3 - 4 | 341 | 55 | |
| NM_001114088.2 | NP_001107560.1 | 3 - 4 | 341 | 55 | ||
| NR_104287.1 | 3 - 4 | 341 | 55 | |||
| GenBank mRNA | AK033091.1 | - | 333 | 55 | ||
| AK049902.1 | - | 3 - 4 | 333 | 55 | ||
| AK077998.1 | - | 3 - 4 | 333 | 55 | ||
| AK128987.1 | - | 3 - 4 | 329 | 55 | ||
| AK142933.1 | - | 3 - 4 | 324 | 55 | ||
| AK167476.1 | - | 3 - 4 | 333 | 55 | ||
| BC052698.1 | - | 3 - 4 | 312 | 55 | ||
More Information
Set Membership: |
|

Pdlim7
Genomic Map