Assay Details
| Gene Symbol: Ecm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012324.2 | NP_001012324.1 | 6 - 7 | 1439 | 104 | |
| GenBank mRNA | AK038812.1 | - | 6 - 7 | 1301 | 104 | |
| AK040995.1 | - | 6 - 7 | 1463 | 104 | ||
| AK134746.1 | - | 6 - 7 | 1393 | 104 | ||
| AK163501.1 | - | 5 - 6 | 3461 | 104 | ||
| BC065151.1 | - | 6 - 7 | 1309 | 5708030 | 104 | |
| BC089559.1 | - | 6 - 7 | 1439 | 30295705 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ecm2
Genomic Map