Assay Details
| Gene Symbol: Ecm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012324.2 | NP_001012324.1 | 7 - 8 | 1597 | 190 | |
| GenBank mRNA | AK038812.1 | - | 7 - 8 | 1459 | 190 | |
| AK040995.1 | - | 7 - 8 | 1621 | 190 | ||
| AK134746.1 | - | 7 - 8 | 1551 | 190 | ||
| AK163501.1 | - | 6 - 7 | 3619 | 190 | ||
| BC065151.1 | - | 7 - 8 | 1467 | 5708030 | 190 | |
| BC089559.1 | - | 7 - 8 | 1597 | 30295705 | 190 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ecm2
Genomic Map