Assay Details
| Gene Symbol: Car9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139305.2 | NP_647466.2 | 6 - 7 | 877 | 79 | |
| GenBank mRNA | AJ245857.1 | - | 6 - 7 | 879 | 79 | |
| AK136579.1 | - | 6 - 7 | 878 | 79 | ||
| AK137719.1 | - | 6 - 7 | 879 | 79 | ||
| AK166403.1 | - | 3 - 4 | 306 | 79 | ||
| BC120544.1 | - | 6 - 7 | 870 | 40129467 | 79 | |
| BC120546.1 | - | 6 - 7 | 870 | 40129469 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car9
Genomic Map