| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400021164.mm.1 |
TC0400002597.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021164.mm.1 |
NM_029086.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021164.mm.1 |
NM_029086.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021161.mm.1 |
TC0400002597.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021161.mm.1 |
NM_029086.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021161.mm.1 |
NM_029086.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021160.mm.1 |
TC0400002597.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021160.mm.1 |
NM_029086.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021160.mm.1 |
NM_029086.2 |
Refseq |