Assay Details
| Gene Symbol: 5830415F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029086.2 | NP_083362.1 | 3 - 4 | 437 | 74 | |
| GenBank mRNA | AK017931.1 | - | 3 - 4 | 448 | 74 | |
| AK044073.1 | - | 3 - 4 | 431 | 74 | ||
| AK052584.1 | - | 3 - 4 | 452 | 74 | ||
| AK153576.1 | - | 3 - 4 | 438 | 74 | ||
| BC092543.1 | - | 3 - 4 | 438 | 30315335 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
acyl-CoA hydrolase activity
biological_process
molecular_function
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
acyl-CoA hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888516 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888517 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164890 | AK153576.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164890 | NM_029086.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164890 | AK044073.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190563 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770343 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672828 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45634 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107185 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112996 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164890 | AK153576.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164890 | NM_029086.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164890 | AK044073.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488768 | AK153576.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488768 | BC092543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488768 | NM_029086.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488768 | AK017931.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P488768 | AK044073.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03742387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03495454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03642258 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03642256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03642255 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03742389 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888516 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888517 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22888515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22888385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03270624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03742389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888504 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04007788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03704298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04007785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03911697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03804812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888507 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03739457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04003469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03638417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03819771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03854635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03891166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03270622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03563201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03746755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03769257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03961095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03495452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03704303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03642250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03854634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03406642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03742387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03460288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03739455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03517107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22888536 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10512754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512755 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512751 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512756 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512752 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512749 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512750 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452873_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512747 | - | - |
| Mouse Genome 430 2.0 Array | 1452873_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452873_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74753_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74753_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400021159.mm.1 | TC0400002597.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021159.mm.1 | NM_029086.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021159.mm.1 | NM_029086.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021157.mm.1 | TC0400002597.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021154.mm.1 | TC0400002597.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021154.mm.1 | NM_029086.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021154.mm.1 | NM_029086.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021158.mm.1 | TC0400002597.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021158.mm.1 | NM_029086.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021158.mm.1 | NM_029086.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021156.mm.1 | TC0400002597.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452873_PM_at | - | - |
| Mouse Expression Set 430 | 1452873_at | - | - |
| Murine Genome U74v2 Set | 167420_PM_at | - | - |
| Murine Genome U74v2 Set | 167420_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452873_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452873_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102470095 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223695 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2924591 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223695 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2924591 | - | - |

5830415F09Rik
Genomic Map