Assay Details
| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023689.3 | NP_076178.1 | 1 - 2 | 188 | 58 | |
| GenBank mRNA | AK013644.1 | - | 1 - 2 | 117 | 58 | |
| AK164338.1 | - | 1 - 2 | 62 | 58 | ||
| AK220280.1 | - | 1 - 2 | 172 | 58 | ||
| BC017601.1 | - | 1 - 2 | 87 | 4502169 | 58 | |
| BC053334.1 | - | 1 - 2 | 59 | 6492113 | 58 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013644.1, AK164338.1, BC017601.1, BC053334.1, NM_023689.3 transcripts.
Set Membership: |
|

Spock3
Genomic Map