Assay Details
| Gene Symbol: Rhobtb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028493.2 | NP_082769.1 | 6 - 7 | 1341 | 75 | |
| GenBank mRNA | AK006650.1 | - | 2 - 3 | 460 | 75 | |
| AK129234.1 | - | 6 - 7 | 1384 | 75 | ||
| AK139158.1 | - | 6 - 7 | 1221 | 75 | ||
| BC050836.1 | - | 6 - 7 | 1341 | 6394317 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhobtb3
Genomic Map