Assay Details
| Gene Symbol: Rhobtb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028493.2 | NP_082769.1 | 7 - 8 | 1449 | 124 | |
| GenBank mRNA | AK006650.1 | - | 3 - 4 | 568 | 124 | |
| AK129234.1 | - | 7 - 8 | 1492 | 124 | ||
| AK139158.1 | - | 7 - 8 | 1329 | 124 | ||
| BC050836.1 | - | 7 - 8 | 1449 | 6394317 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhobtb3
Genomic Map