Assay Details
| Gene Symbol: 2310067P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010100.1 | - | 2 - 3 | 523 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3961 | Mir3961 | mmu-mir-3961 | -367.9kb | Anti-sense | Chr. 13 - 82698275 - 82698333 on NCBI Build GRCm38 |
| Downstream | myocyte enhancer factor 2C | Mef2c |
5430401D19Rik, 9930028G15Rik, AV011172, Mef2 |
408.2kb | Sense | Chr. 13 - 83504034 - 83667079 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

2310067P03Rik
Genomic Map