Assay Details
| Gene Symbol: Dzip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110017.1 | NP_001103487.1 | 8 - 9 | 1084 | 131 | |
| NM_027341.2 | NP_081617.1 | 8 - 9 | 1084 | 131 | ||
| GenBank mRNA | AK039172.1 | - | 8 - 9 | 1062 | 131 | |
| BC052893.1 | - | 8 - 9 | 892 | 131 | ||
| BC117953.1 | - | 8 - 9 | 907 | 40098141 | 131 | |
| BC117954.1 | - | 8 - 9 | 913 | 40098143 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dzip3
Genomic Map