Assay Details
| Gene Symbol: Golgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030035.1 | NP_084311.1 | 11 - 12 | 1704 | 132 | |
| GenBank mRNA | BC145221.1 | - | 11 - 12 | 1663 | 9053772 | 132 |
| BC150731.1 | - | 11 - 12 | 1681 | 9087642 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golgb1
Genomic Map