Assay Details
| Gene Symbol: Golgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030035.1 | NP_084311.1 | 17 - 18 | 9286 | 113 | |
| GenBank mRNA | AK220188.1 | - | 5 - 6 | 5141 | 113 | |
| BC040461.1 | - | 5 - 6 | 2606 | 113 | ||
| BC042759.1 | - | 4 - 5 | 621 | 113 | ||
| BC060254.1 | - | 4 - 5 | 1710 | 113 | ||
| BC145221.1 | - | 15 - 16 | 2228 | 9053772 | 113 | |
| BC150731.1 | - | 17 - 18 | 9263 | 9087642 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golgb1
Genomic Map