| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600004977.mm.1 |
NM_030035.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004977.mm.1 |
NM_030035.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004977.mm.1 |
TC1600000528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004975.mm.1 |
NM_030035.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004975.mm.1 |
NM_030035.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004975.mm.1 |
TC1600000528.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600004976.mm.1 |
NM_030035.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004976.mm.1 |
NM_030035.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600004976.mm.1 |
TC1600000528.mm.1 |
MTA TC |