Assay Details
| Gene Symbol: Olfr183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146485.2 | NP_666696.2 | 2 | 314 | 72 | |
| GenBank mRNA | DR065568.1 | - | 2 | 333 | 72 | |
| DR065627.1 | - | 2 | 135 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr183
Genomic Map