Assay Details
| Gene Symbol: Dgkg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138650.2 | NP_619591.1 | 3 - 4 | 604 | 66 | |
| GenBank mRNA | AK083258.1 | - | 3 - 4 | 597 | 66 | |
| AK087972.1 | - | 3 - 4 | 562 | 66 | ||
| AK089261.1 | - | 3 - 4 | 402 | 66 | ||
| AK220294.1 | - | 3 - 4 | 546 | 66 | ||
| BC015278.1 | - | 2 - 3 | 177 | 4221290 | 66 | |
| BY724037.1 | - | 3 - 4 | 604 | 66 | ||
More Information
Set Membership: |
|

Dgkg
Genomic Map