Assay Details
| Gene Symbol: Bphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026512.1 | NP_080788.1 | 3 - 4 | 403 | 85 | |
| GenBank mRNA | AK002907.1 | - | 3 - 4 | 401 | 85 | |
| AK007606.1 | - | 3 - 4 | 397 | 85 | ||
| AK017801.1 | - | 3 - 4 | 406 | 85 | ||
| AK078232.1 | - | 3 - 4 | 448 | 85 | ||
| AK152712.1 | - | 3 - 4 | 413 | 85 | ||
| AK170099.1 | - | 3 - 4 | 406 | 85 | ||
| BC023146.1 | - | 3 - 4 | 388 | 3990118 | 85 | |
| BC025162.1 | - | 3 - 4 | 403 | 4921451 | 85 | |
| BC029322.1 | - | 1 - 2 | 38 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bphl
Genomic Map