Assay Details
| Gene Symbol: Fam193b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145382.4 | NP_663357.2 | 9 - 10 | 3844 | 80 | |
| GenBank mRNA | AK040568.1 | - | 3 - 4 | 1972 | 80 | |
| AK155237.1 | - | 9 - 10 | 3838 | 80 | ||
| AK173303.1 | - | 9 - 10 | 2693 | 80 | ||
| BC021381.1 | - | 3 - 4 | 1434 | 5043282 | 80 | |
| BC023015.1 | - | 3 - 4 | 1376 | 5364758 | 80 | |
| BC025483.1 | - | 9 - 10 | 2703 | 5150988 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam193b
Genomic Map