Assay Details
| Gene Symbol: Pdia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028295.1 | NP_082571.1 | 1 - 2 | 142 | 95 | |
| GenBank mRNA | BC009151.1 | - | 1 - 2 | 142 | 3600849 | 95 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC009151.1, NM_028295.1 transcripts.
Set Membership: |
|

Pdia5
Genomic Map