Assay Details
| Gene Symbol: Cenpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025495.3 | NP_079771.1 | 4 - 5 | 500 | 126 | |
| GenBank mRNA | AK006234.1 | - | 4 - 5 | 537 | 126 | |
| AK012863.1 | - | 4 - 5 | 504 | 126 | ||
| AK014919.1 | - | 4 - 5 | 456 | 126 | ||
| BC094280.1 | - | 4 - 5 | 553 | 126 | ||
| BC147207.1 | - | 4 - 5 | 480 | 9056262 | 126 | |
| BC147208.1 | - | 4 - 5 | 480 | 9056260 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpp
Genomic Map