Assay Details
| Gene Symbol: Tctex1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025329.3 | NP_079605.1 | 4 - 5 | 487 | 64 | |
| GenBank mRNA | AK002589.1 | - | 4 - 5 | 446 | 64 | |
| AK004191.1 | - | 3 - 4 | 345 | 64 | ||
| AK007178.1 | - | 4 - 5 | 475 | 64 | ||
| AK010027.1 | - | 4 - 5 | 445 | 64 | ||
| AK010464.1 | - | 3 - 4 | 373 | 64 | ||
| BC053332.1 | - | 2 - 3 | 1807 | 64 | ||
| BC059716.1 | - | 4 - 5 | 488 | 6774264 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tctex1d2
Genomic Map