Assay Details
| Gene Symbol: Melk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010790.2 | NP_034920.2 | 11 - 12 | 1112 | 58 | |
| GenBank mRNA | AK010498.1 | - | 11 - 12 | 1078 | 58 | |
| AK011932.1 | - | 11 - 12 | 1112 | 58 | ||
| AK129076.1 | - | 11 - 12 | 1065 | 58 | ||
| AK137000.1 | - | 11 - 12 | 1138 | 58 | ||
| AK145021.1 | - | 11 - 12 | 1047 | 58 | ||
| AK145316.1 | - | 11 - 12 | 1021 | 58 | ||
| AK161029.1 | - | 11 - 12 | 1121 | 58 | ||
| AK164138.1 | - | 11 - 12 | 1079 | 58 | ||
| AK167457.1 | - | 11 - 12 | 1036 | 58 | ||
| BC020416.1 | - | 10 - 11 | 947 | 58 | ||
| L76158.1 | - | 11 - 12 | 1043 | 58 | ||
| X95351.1 | - | 11 - 12 | 1050 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Melk
Genomic Map