Assay Details
| Gene Symbol: Melk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010790.2 | NP_034920.2 | 14 - 15 | 1584 | 79 | |
| GenBank mRNA | AK010498.1 | - | 14 - 15 | 1550 | 79 | |
| AK011932.1 | - | 14 - 15 | 1584 | 79 | ||
| AK129076.1 | - | 14 - 15 | 1537 | 79 | ||
| AK137000.1 | - | 14 - 15 | 1610 | 79 | ||
| AK145021.1 | - | 14 - 15 | 1519 | 79 | ||
| AK145316.1 | - | 14 - 15 | 1493 | 79 | ||
| AK161029.1 | - | 14 - 15 | 1593 | 79 | ||
| AK164138.1 | - | 14 - 15 | 1551 | 79 | ||
| AK167457.1 | - | 14 - 15 | 1508 | 79 | ||
| BC020416.1 | - | 13 - 14 | 1419 | 79 | ||
| BC085276.1 | - | 14 - 15 | 1617 | 30443236 | 79 | |
| L76158.1 | - | 14 - 15 | 1515 | 79 | ||
| X95351.1 | - | 14 - 15 | 1522 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Melk
Genomic Map