Assay Details
| Gene Symbol: Melk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010790.2 | NP_034920.2 | 6 - 7 | 668 | 113 | |
| GenBank mRNA | AK010498.1 | - | 6 - 7 | 634 | 113 | |
| AK011932.1 | - | 6 - 7 | 668 | 113 | ||
| AK129076.1 | - | 6 - 7 | 621 | 113 | ||
| AK137000.1 | - | 6 - 7 | 694 | 113 | ||
| AK145021.1 | - | 6 - 7 | 603 | 113 | ||
| AK145316.1 | - | 6 - 7 | 577 | 113 | ||
| AK161029.1 | - | 6 - 7 | 677 | 113 | ||
| AK164138.1 | - | 6 - 7 | 635 | 113 | ||
| AK167457.1 | - | 6 - 7 | 592 | 113 | ||
| BC020416.1 | - | 5 - 6 | 503 | 113 | ||
| BC085276.1 | - | 6 - 7 | 701 | 30443236 | 113 | |
| L76158.1 | - | 6 - 7 | 599 | 113 | ||
| X95351.1 | - | 6 - 7 | 606 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Melk
Genomic Map