Assay Details
| Gene Symbol: Otud3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028453.1 | NP_082729.1 | 3 - 4 | 595 | 66 | |
| GenBank mRNA | AK014098.1 | - | 3 - 4 | 637 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014098.1 transcripts.
Set Membership: |
|

Otud3
Genomic Map