Assay Details
| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284375.1 | NP_001271304.1 | 11 - 12 | 1515 | 66 | |
| NM_175650.4 | NP_783581.2 | 12 - 13 | 1650 | 66 | ||
| GenBank mRNA | AK083102.1 | - | 12 - 13 | 1625 | 66 | |
| AK083121.1 | - | 12 - 13 | 1651 | 66 | ||
| AK158355.1 | - | 12 - 13 | 1470 | 66 | ||
| BC115739.1 | - | 11 - 12 | 1448 | 40086065 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp13a5
Genomic Map