Assay Details
| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284375.1 | NP_001271304.1 | 18 - 19 | 2385 | 100 | |
| NM_175650.4 | NP_783581.2 | 19 - 20 | 2520 | 100 | ||
| GenBank mRNA | AK083102.1 | - | 19 - 20 | 2495 | 100 | |
| AK083121.1 | - | 19 - 20 | 2521 | 100 | ||
| AK158355.1 | - | 19 - 20 | 2340 | 100 | ||
| BC115739.1 | - | 18 - 19 | 2318 | 40086065 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp13a5
Genomic Map