Assay Details
| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284375.1 | NP_001271304.1 | 25 - 26 | 3074 | 77 | |
| NM_175650.4 | NP_783581.2 | 26 - 27 | 3209 | 77 | ||
| GenBank mRNA | AK083102.1 | - | 27 - 28 | 3247 | 77 | |
| AK083121.1 | - | 26 - 27 | 3210 | 77 | ||
| AK158355.1 | - | 26 - 27 | 3029 | 77 | ||
| BC115739.1 | - | 25 - 26 | 3007 | 40086065 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp13a5
Genomic Map