Assay Details
| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284375.1 | NP_001271304.1 | 4 - 5 | 654 | 77 | |
| NM_175650.4 | NP_783581.2 | 4 - 5 | 654 | 77 | ||
| GenBank mRNA | AK083102.1 | - | 4 - 5 | 629 | 77 | |
| AK083121.1 | - | 4 - 5 | 655 | 77 | ||
| AK158355.1 | - | 4 - 5 | 474 | 77 | ||
| CB524474.1 | - | 4 - 5 | 456 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp13a5
Genomic Map