Assay Details
| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284375.1 | NP_001271304.1 | 7 - 8 | 879 | 127 | |
| NM_175650.4 | NP_783581.2 | 8 - 9 | 1014 | 127 | ||
| GenBank mRNA | AK083102.1 | - | 8 - 9 | 989 | 127 | |
| AK083121.1 | - | 8 - 9 | 1015 | 127 | ||
| AK158355.1 | - | 8 - 9 | 834 | 127 | ||
| BC115739.1 | - | 7 - 8 | 812 | 40086065 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC115739.1 transcripts.
Set Membership: |
|

Atp13a5
Genomic Map