Assay Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025608.2 | NP_001020779.1 | 1 - 2 | 128 | ||
| NM_001277195.1 | NP_001264124.1 | 1 - 2 | 125 | |||
| NR_102344.1 | 1 - 2 | 99 | 75 | |||
| GenBank mRNA | BC066992.1 | - | 1 - 2 | 5685985 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Szrd1
Genomic Map