Assay Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025608.2 | NP_001020779.1 | 2 - 3 | 158 | 83 | |
| GenBank mRNA | BC066992.1 | - | 2 - 3 | 106 | 5685985 | 83 |
More Information
Set Membership: |
|

Szrd1
Genomic Map