Assay Details
| Gene Symbol: C330004M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021176.1 | - | 1 - 2 | 863 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | xyloside xylosyltransferase 1 | Xxylt1 | AI480653 | Sense | Chr. 16 - 30928348 - 31081551 on NCBI Build GRCm38 | |
| Downstream |
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2 |
Acap2 |
4832442G16, 9530039J15Rik, CNT-B2, Centb2, mKIAA0041 |
156.2kb | Sense | Chr. 16 - 31092412 - 31202056 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

C330004M20Rik
Genomic Map