Assay Details
Gene Symbol: Gm17750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK082072.1 | - | 1 - 2 | 126 | 81 | |
BE862222.1 | - | 1 - 2 | 128 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 2810049E08 gene | 2810049E08Rik | -96.6kb | Anti-sense | Chr. 13 - 83891208 - 83928710 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA A230107N01 gene | A230107N01Rik | 154.9kb | Sense | Chr. 13 - 84219580 - 84222230 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282996 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535528 | AF541936.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535528 | BC052041.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 535528 | NM_177360.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 311136 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355133 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295348 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523982 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712132 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484428 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953997 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487590 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808620 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77743 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134114 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61824 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07938596 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07938551 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07938569 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103709 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07938596 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189319 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189263 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189004 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189327 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11115598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189269 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10664989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189287 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189329 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189280 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189264 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189128 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29188997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189063 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189039 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189102 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10681751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189043 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189057 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189061 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189002 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10915176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10763217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189100 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10588133 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189040 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189000 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189042 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189310 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189323 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189027 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10815430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189309 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189341 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29188998 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189324 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10588140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189116 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189313 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189025 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189270 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189058 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10815439 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189005 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189121 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189045 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10811362 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189122 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189336 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189333 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189321 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189064 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189103 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29188996 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29188999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189275 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189350 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189353 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189059 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189001 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189331 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189283 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189029 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29189277 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1455557_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455557_at | - | - |
Mouse Genome 430 2.0 Array | 1455557_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300015794.mm.1 | TC1300002373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300015789.mm.1 | TC1300002373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300015787.mm.1 | TC1300002373.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300015788.mm.1 | TC1300002373.mm.1 | MTA TC |
Mouse Expression Set 430 | 1455557_at | - | - |
Mouse Expression Set 430 | 1455557_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455557_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455557_PM_at | - | - |
