Assay Details
| Gene Symbol: Ctnnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165902.1 | NP_001159374.1 | 6 - 7 | 1019 | 69 | |
| NM_007614.3 | NP_031640.1 | 6 - 7 | 1219 | 69 | ||
| GenBank mRNA | AK018515.1 | - | 6 - 7 | 1133 | 69 | |
| AK020013.1 | - | 6 - 7 | 1135 | 69 | ||
| AK035311.1 | - | 6 - 7 | 1132 | 69 | ||
| AK132398.1 | - | 6 - 7 | 1200 | 69 | ||
| AK133656.1 | - | 6 - 7 | 1132 | 69 | ||
| AK135743.1 | - | 3 - 4 | 686 | 69 | ||
| AK211747.1 | - | 1 - 2 | 63 | 69 | ||
| AY751549.1 | - | 6 - 7 | 1144 | 69 | ||
| BC043078.1 | - | 3 - 4 | 621 | 69 | ||
| BC043481.1 | - | 2 - 3 | 253 | 69 | ||
| BC048153.1 | - | 6 - 7 | 1144 | 5709247 | 69 | |
| BC053065.1 | - | 6 - 7 | 1144 | 5709247 | 69 | |
| M90364.1 | - | 6 - 7 | 1029 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnb1
Genomic Map