Assay Details
| Gene Symbol: Ctnnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165902.1 | NP_001159374.1 | 9 - 10 | 1612 | 77 | |
| NM_007614.3 | NP_031640.1 | 9 - 10 | 1812 | 77 | ||
| GenBank mRNA | AK018515.1 | - | 9 - 10 | 1726 | 77 | |
| AK020013.1 | - | 9 - 10 | 1728 | 77 | ||
| AK035311.1 | - | 9 - 10 | 1725 | 77 | ||
| AK132398.1 | - | 9 - 10 | 1793 | 77 | ||
| AK133656.1 | - | 9 - 10 | 1725 | 77 | ||
| AK135743.1 | - | 6 - 7 | 1279 | 77 | ||
| BC043078.1 | - | 6 - 7 | 1214 | 77 | ||
| BC043481.1 | - | 5 - 6 | 846 | 77 | ||
| BC048153.1 | - | 9 - 10 | 1737 | 5709247 | 77 | |
| BC053065.1 | - | 9 - 10 | 1737 | 5709247 | 77 | |
| M90364.1 | - | 9 - 10 | 1622 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnb1
Genomic Map