Assay Details
| Gene Symbol: Ctnnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165902.1 | NP_001159374.1 | 10 - 11 | 1772 | 88 | |
| NM_007614.3 | NP_031640.1 | 10 - 11 | 1972 | 88 | ||
| GenBank mRNA | AK018515.1 | - | 10 - 11 | 1886 | 88 | |
| AK035311.1 | - | 10 - 11 | 1885 | 88 | ||
| AK132398.1 | - | 10 - 11 | 1953 | 88 | ||
| AK133656.1 | - | 10 - 11 | 1885 | 88 | ||
| AK135743.1 | - | 7 - 8 | 1439 | 88 | ||
| BC043078.1 | - | 7 - 8 | 1374 | 88 | ||
| BC043481.1 | - | 6 - 7 | 1006 | 88 | ||
| BC048153.1 | - | 10 - 11 | 1897 | 5709247 | 88 | |
| BC053065.1 | - | 10 - 11 | 1897 | 5709247 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnb1
Genomic Map