Assay Details
| Gene Symbol: Brd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286630.1 | NP_001273559.1 | 1 - 2 | 69 | 122 | |
| NM_020508.4 | NP_065254.3 | 1 - 2 | 69 | 122 | ||
| NM_198094.2 | NP_932762.2 | 1 - 2 | 59 | 122 | ||
| GenBank mRNA | AF461395.1 | - | 1 - 2 | 45 | 122 | |
| AF461396.1 | - | 1 - 2 | 45 | 122 | ||
| BC138834.1 | - | 1 - 2 | 39 | 8861856 | 122 | |
| BC138835.1 | - | 1 - 2 | 38 | 8861857 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF461395.1, AF461396.1, BC138834.1, BC138835.1, NM_001286630.1, NM_020508.4, NM_198094.2 transcripts.
Set Membership: |
|

Brd4
Genomic Map