Assay Details
| Gene Symbol: Cmtm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027294.2 | NP_081570.1 | 1 - 2 | 372 | 97 | |
| GenBank mRNA | AK012262.1 | - | 1 - 2 | 373 | 97 | |
| AY243513.1 | - | 1 - 2 | 407 | 97 | ||
| BC027554.1 | - | 1 - 2 | 294 | 1382818 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm8
Genomic Map