Assay Details
| Gene Symbol: Agmat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081408.1 | NP_001074877.1 | 5 - 6 | 1047 | 63 | |
| GenBank mRNA | AK017155.1 | - | 5 - 6 | 944 | 63 | |
| BC025916.1 | - | 2 - 3 | 264 | 63 | ||
| BC037459.1 | - | 2 - 3 | 220 | 63 | ||
| BC115693.2 | - | 5 - 6 | 944 | 40104399 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Agmat
Genomic Map