Assay Details
| Gene Symbol: Lipi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252513.1 | NP_001239442.1 | 3 - 4 | 593 | 115 | |
| GenBank mRNA | AK086576.1 | - | 3 - 4 | 596 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lipi
Genomic Map