Assay Details
| Gene Symbol: Galnt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172693.3 | NP_766281.1 | 8 - 9 | 1456 | 80 | |
| GenBank mRNA | AB175680.1 | - | 8 - 9 | 1436 | 80 | |
| AK033638.1 | - | 8 - 9 | 1457 | 80 | ||
| AK042133.1 | - | 8 - 9 | 1489 | 80 | ||
| AK043977.1 | - | 7 - 8 | 1317 | 80 | ||
| AK082014.1 | - | 8 - 9 | 1502 | 80 | ||
| AK156751.1 | - | 8 - 9 | 1486 | 80 | ||
| BC024475.1 | - | 7 - 8 | 1288 | 80 | ||
| BC025639.1 | - | 7 - 8 | 1178 | 80 | ||
| BC056425.1 | - | 8 - 9 | 1467 | 5716909 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt12
Genomic Map