Assay Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081154.2 | NP_001074623.1 | 1 - 2 | 128 | 101 | |
| GenBank mRNA | AK220341.1 | - | 1 - 2 | 81 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001081154.2 transcripts.
Set Membership: |
|

Marf1
Genomic Map